Orders LinkContactUsLinkComponent
Connector kits, SeCure® 'Y'
Connector kits, SeCure® 'Y'
Supplier:   Restek
Connector kits, SeCure® 'Y'
Supplier:  Restek
Certificates
Connector kits, SeCure® 'Y'
Connector kits, SeCure® 'Y'
4 Options Available Below
Product Details & Documents
Connector kits connects two analytical columns to a transfer line or guard column. They have a reliable seal integrity and will not unexpectedly disconnect during temperature programmed analyses. The open design allows visual confirmation of the seal for added confidence in the connection.
Combine the 'Y' Press-Tight® connector with the strength of a metal union. The ferrules and knurled nuts hold the fused silica tubing in place, which prevents the tubing from unexpectedly disconnecting, even at temperatures as high as 400 °C.
Delivery information: Kits include: SeCure® 'Y' connector body, three knurled nuts, universal 'Y' Press-Tight® union and three ferrules.
Product Family Options
Product Information
  • Availability
    Price
  • REST20276
    Connector kit for 0,18/0,25/0,28 mm column ID
    Connector kit for 0,18/0,25/0,28 mm column ID
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 421,00 €
    421,00 €
    1 * 1 ST
       421,00 €
  • REST20277
    Connector kit for 0,32 mm column ID
    Connector kit for 0,32 mm column ID
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 421,00 €
    421,00 €
    1 * 1 ST
       421,00 €
  • REST20278
    Connector kit for 0,45/0,53 mm column ID
    Connector kit for 0,45/0,53 mm column ID
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 421,00 €
    421,00 €
    1 * 1 ST
       421,00 €
  • REST20279
    Knurled nut
    Knurled nut
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST