Orders LinkContactUsLinkComponent
Diffraction/transmission gratings
Diffraction/transmission gratings
Marketsource
Supplier:   3B Scientific
Diffraction/transmission gratings
Supplier:  3B Scientific
Marketsource
Certificates
Diffraction/transmission gratings
Diffraction/transmission gratings
2 Options Available Below
Product Details & Documents
Transmission grating for spectroscopic examinations and for experiments on diffraction and interference. Suitable to resolve the sodium D lines.
  • Mounted on glass carrier
  • Dimensions: 38×50 mm
Product Family Options
Product Information
  • Availability
    Price
  • BINH1003079
    Transmission grating 600 lines/mm
    Transmission grating 600 lines/mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 291,00 €
    291,00 €
    1 * 1 ST
       291,00 €
    Specifications:
    Marketsource
    Marketsource
  • BINH1003080
    Transmission grating 300 lines/mm
    Transmission grating 300 lines/mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 244,00 €
    244,00 €
    1 * 1 ST
       244,00 €
    Specifications:
    Marketsource
    Marketsource