Reagent bottles, with screw caps
Catalog # 1.13314.0001
Supplier: Merck
Specifications
- Thread:S 40
- Capacity:1000 ml
- Description:Bottles, glass, PE coated
- Seq:06
- Environmentally Preferable:
- Pk:6
Specifications
About this item
Glass, amber.
Amber coloured to protect light-sensitive materials.