Order Entry
Austria
ContactUsLinkComponent
pH/mV/°C meters, bench, FiveEasy™ F20 / FiveEasy™ Plus FP20
pH/mV/°C meters, bench, FiveEasy™ F20 / FiveEasy™ Plus FP20
Supplier:  Mettler - Toledo
Certificates
undefined
pH/mV/°C meters, bench, FiveEasy™ F20 / FiveEasy™ Plus FP20
Supplier:  Mettler - Toledo
Certificates

Specifications

  • Catalog No:
  • 662-2002
  • 662-2003
  • 662-2011
  • 662-2004
  • 662-2005
  • 662-2001
  • 662-2006
  • Model:
  • FP20
  • FP20
  • F20
  • FP20
  • FP20
  • F20
  • FP20
  • pH range:
  • −2,00...+16,00
  • −2,00...+16,00
  • 0,00...+14,00
  • −2,00...+16,00
  • −2,00...+16,00
  • 0,00...+14,00
  • −2,00...+16,00
  • pH resolution:
  • 0,01/0,1
  • 0,01/0,1
  • 0,01
  • 0,01/0,1
  • 0,01/0,1
  • 0,01
  • 0,01/0,1
  • pH accuracy:
  • ±0,01
  • ±0,01
  • ±0,01
  • ±0,01
  • ±0,01
  • ±0,01
  • ±0,01
  • pH calibration:
  • Linear/segmented, max. 5 points (4 pre-defined buffer groups)
  • Linear/segmented, max. 5 points (4 pre-defined buffer groups)
  • Linear, max. 3 points (4 pre-defined buffer groups)
  • Linear/segmented, max. 5 points (4 pre-defined buffer groups)
  • Linear/segmented, max. 5 points (4 pre-defined buffer groups)
  • Linear, max. 3 points (4 pre-defined buffer groups)
  • Linear/segmented, max. 5 points (4 pre-defined buffer groups)
  • mV range:
  • ±2000
  • ±2000
  • ±2000
  • ±2000
  • ±2000
  • ±2000
  • ±2000
  • mV resolution:
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • mV accuracy:
  • ±1
  • ±1
  • ±1
  • ±1
  • ±1
  • ±1
  • ±1
  • Temperature range:
  • −5...+105 °C
  • −5...+105 °C
  • 0...+100 °C
  • −5...+105 °C
  • −5...+105 °C
  • 0...+100 °C
  • −5...+105 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,3 °C
  • ±0,3 °C
  • ±0,5 °C
  • ±0,3 °C
  • ±0,3 °C
  • ±0,5 °C
  • ±0,3 °C
  • Temperature compensation:
  • ATC & MTC
  • ATC & MTC
  • ATC & MTC
  • ATC & MTC
  • ATC & MTC
  • ATC & MTC
  • ATC & MTC
  • Display:
  • Segmented LCD
  • Segmented LCD
  • Segmented LCD
  • Segmented LCD
  • Segmented LCD
  • Segmented LCD
  • Segmented LCD
  • Power supply:
  • 100 - 240 V; 50 - 60 Hz
  • 100 - 240 V; 50 - 60 Hz
  • 100 - 240 V; 50 - 60 Hz
  • 100 - 240 V; 50 - 60 Hz
  • 100 - 240 V; 50 - 60 Hz
  • 100 - 240 V; 50 - 60 Hz
  • 100 - 240 V; 50 - 60 Hz
  • Data logging capacity:
  • 200 measurements
  • 200 measurements
  • -
  • 200 measurements
  • 200 measurements
  • -
  • 200 measurements
  • Outputs:
  • USB, RS232
  • USB, RS232
  • Analog
  • USB, RS232
  • USB, RS232
  • Analog
  • USB, RS232
  • W×D×H:
  • 227×147×70 mm
  • 227×147×70 mm
  • 227×147×70 mm
  • 227×147×70 mm
  • 227×147×70 mm
  • 227×147×70 mm
  • 227×147×70 mm
  • Weight:
  • 0,63 kg
  • 0,63 kg
  • 0,63 kg
  • 0,63 kg
  • 0,63 kg
  • 0,63 kg
  • 0,63 kg

Specifications

Related Information

Certificates
Search by Lot Number

Electrodes

Product Information

Frequently Bought Together