Orders LinkContactUsLinkComponent
VWR® Buffer Standard Solutions
VWR® Buffer Standard Solutions
VWR® Buffer Standard Solutions
Supplier:  VWR Chemicals
Certificates
VWR® Buffer Standard Solutions
VWR® Buffer Standard Solutions
3 Options Available Below
Product Family Options
Product Information
  • pH value
    pH accuracy
    Reference temperature
    Pack type
    Pk
    Availability
    Price
  • 4,00
    ±0,03
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 69,50 €
    69,50 €
    1 * 500 mL
       69,50 €
  • 7,00
    ±0,03
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 78,40 €
    78,40 €
    1 * 500 mL
       78,40 €
  • 10,0
    ±0,03
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 96,90 €
    96,90 €
    1 * 500 mL
       96,90 €