pH combination electrodes, IDS®
Supplier: SI Analytics
Jump to:
About this item
When digital IDS sensors are connected to the measuring device, they automatically identify themselves with their serial numbers and type designation and transmit their calibration data to the device.
- Calibration data is stored in the sensor head
- Measuring signals are transmitted without interference
- Perfect galvanized separation
- Resistant against environmental influences
- Allows prognostic maintenance of the sensors by the intelligent sensor evaluation QSC
- Effortless allocation and documentation of the sensor to electronically captured and saved calibration results
- Highest possible operator comfort and measuring accuracy
3 Options Available Below
Product Details & Documents
When digital IDS sensors are connected to the measuring device, they automatically identify themselves with their serial numbers and type designation and transmit their calibration data to the device.
- Calibration data is stored in the sensor head
- Measuring signals are transmitted without interference
- Perfect galvanized separation
- Resistant against environmental influences
- Allows prognostic maintenance of the sensors by the intelligent sensor evaluation QSC
- Effortless allocation and documentation of the sensor to electronically captured and saved calibration results
- Highest possible operator comfort and measuring accuracy
Product Family Options
Product Information
- TypepHTemp.MaterialElectrolyteDiaphr.Ø×LConnectionForAvailabilityPrice
- SCOI285100080A 157 IDS®, micro electrode with NTC 30 kΩA 157 IDS®, micro electrode with NTC 30 kΩA 157 IDS®, micro electrode0 - 14−5...+100 °CGlassKCl 3 mol/lPlatinum12/5×200 mmIDS® plug, 1,5 m cableLife Science applicationsLog in for availability1 * 1 ST669,00 €
- 661-0244A 162 IDS®, with NTC 30 kΩA 162 IDS®, with NTC 30 kΩA 162 IDS®0 - 14−5...+100 °CGlassKCl 3 mol/lPlatinum12×120 mmIDS® plug, 1,5 m cableDemanding applicationsLog in for availability1 * 1 ST431,00 €
- SCOI285101080A 7780 IDS®, with NTC 30 kΩA 7780 IDS®, with NTC 30 kΩA 7780 IDS®0 - 14−5...+80 °CGlassGel3 × ceramic12×120 mmIDS® plug, 1,5 m cableVersatile applicationsLog in for availability1 * 1 ST387,00 €