Vacuum aspiration systems, AE and AK
The simple vacuum safety aspiration system, together with the quick lock adapters, filter, ventilation insert and the silicone tubing forms a complete unit. The comfortable aspiration devices additionally have a bottle holder and handle.
- System optionally available with 2 or 4-litre bottle
- Screw cap for the bottles with two quick action couplings that close automatically if the hose connections are disengaged
- Extra-long (3 m) silicone tubing allows the bottle and the vacuum pump to be placed away from the workbench
Frequently Bought Together