Density hydrometers, M100
Supplier: GERING
Certificates
About this item
Reference temperature for density measurements: 20 °C.
Certifications: DIN 12791
Packaging: In velvet lined case.
Frequently Bought Together
Reference temperature for density measurements: 20 °C.