Stereo microscopes, Stemi 508
Supplier: ZEISS
About this item
The Stemi 508 are designed for quality images in biomedical (IVF), botany (plants, insects) and forensics (dissection, fossiles) applications.
- Large object field for details (36 mm)
- Contrast and colour reproducibility
- Robust design with camera output for outstanding three dimensional images
Ordering information: Eyepieces are included.
Delivery information: Supplied with a VisiLED ring light.
3 Options Available Below
Product Details & Documents
The Stemi 508 are designed for quality images in biomedical (IVF), botany (plants, insects) and forensics (dissection, fossiles) applications.
- Large object field for details (36 mm)
- Contrast and colour reproducibility
- Robust design with camera output for outstanding three dimensional images
Ordering information: Eyepieces are included.
Delivery information: Supplied with a VisiLED ring light.
Specifications for Product Family
Specifications
- Catalog No:
- 630-2822
- 630-2821
- 630-2632
- Model:
- Stemi 508 with segmentable LED ring
- Stemi 508 Doc
- Stemi 508 Edu
- Eyepieces:
- WF 10×/23 mm
- WF 10×/23 mm
- WF 10×/23 mm
- Magnification:
- 13× - 100×
- 6,5× - 50×
- 13× - 100×
- Zoom lens:
- 8:1
- 8:1
- 8:1
- Focusing system:
- Separated coarse and fine knobs
- Separated coarse and fine knobs
- Separated coarse and fine knobs
- Illumination:
- LED
- LED
- LED
Specifications
Product Family Options
Product Information
- ModelTypeAvailabilityPrice
- 630-2822Stereo microscopeSpecifications:
More
Specifications
Cat. No.630-2822ModelStemi 508 with segmentable LED ringEyepiecesWF 10×/23 mmMagnification13× - 100×Zoom lens8:1Focusing systemSeparated coarse and fine knobsIlluminationLED - 630-2821Stereo microscopeSpecifications:
More
Specifications
Cat. No.630-2821ModelStemi 508 DocEyepiecesWF 10×/23 mmMagnification6,5× - 50×Zoom lens8:1Focusing systemSeparated coarse and fine knobsIlluminationLED - 630-2632Stereo microscopeSpecifications:
More
Specifications
Cat. No.630-2632ModelStemi 508 EduEyepiecesWF 10×/23 mmMagnification13× - 100×Zoom lens8:1Focusing systemSeparated coarse and fine knobsIlluminationLED
Accessories
Product Information
- Catalog #AvailabilityPrice
- Transport case