Anti-CD3,CD11b/c,CD19,CD45R/B220,Ly-6G (Gr-1),TER-119 Monoclonal Antibody (redFluor®710, violetFluor®450,PerCP-Cy5.5®,PE-Cy7®,FITC) [clone:53-6.7,M1/70,N418,RA3-6B2,RB6-8C5,M5/114.15.2]
Catalog # 91-0003-U025
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Primary
- Antigen name:CD3 Molecule
- Antigen symbol:CD3
- Clonality:Monoclonal
- Clone:53-6.7, M1/70, N418, RA3-6B2, RB6-8C5, M5/114.15.2
- Conjugation:redFluor® 710/violetFluor® 450/PerCP-Cy5.5®/PE-Cy7®/FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Multiple
- Isotype:
- Reactivity:Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Antigen synonyms:CD11c|CD3|Ly-6G (Gr-1)|CD45R/B220|CD11b|CD19|TER-119
- Storage temperature:Store undiluted at 2-8 °C, protect from light, do not freeze.
- Size:25 μg
- Pk:25 µG
Specifications
About this item
The Mouse Myeloid Precursor backbone panel is a five-color combination of seven anti-mouse antibodies. This panel can serve as a backbone to identify various myeloid precursor populations, and leaves room for GFP or additional drop-in markers, if desired. The CD3e, CD19 and CD45R/B220 positive cells are gated together to exclude mature T and B cells. Violet laser (405 nm) required for detection of CD11b violetFluorTM 450. Hosts: Rat, Armenian Hamster
Type: Primary
Antigen: CD3, CD11b, CD11c, CD19, CD45R/B220, Ly-6G (Gr-1), TER-119
Clonality: Monoclonal
Clone: 53-6.7, M1/70, N418, RA3-6B2, RB6-8C5, M5/114.15.2
Conjugation: redFluor® 710, violetFluor® 450, PerCP-Cy5.5®, PE-Cy7®, FITC
Epitope:
Host: Rat, Armenian Hamster
Isotype:
Reactivity: Mouse