Order Entry
Austria
ContactUsLinkComponent
Anti-CD3, CD4, CD8a, CD25, CD44, CD62L (L-Selectin) Rat Monoclonal Antibody (violetFluor®450,APC-Cy7®,redFluor®710,PE-Cy7®,PerCP-Cy5.5®,APC) [clone:17A2,RM4-5, 53-6.7,PC61.5,IM7,MEL-14]
Anti-CD3, CD4, CD8a, CD25, CD44, CD62L (L-Selectin) Rat Monoclonal Antibody (violetFluor®450,APC-Cy7®,redFluor®710,PE-Cy7®,PerCP-Cy5.5®,APC) [clone:17A2,RM4-5, 53-6.7,PC61.5,IM7,MEL-14]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD3, CD4, CD8a, CD25, CD44, CD62L (L-Selectin) Rat Monoclonal Antibody (violetFluor®450,APC-Cy7®,redFluor®710,PE-Cy7®,PerCP-Cy5.5®,APC) [clone:17A2,RM4-5, 53-6.7,PC61.5,IM7,MEL-14]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 91-0002-U025
  • 91-0002-U100
  • Antigen symbol:
  • CD3
  • CD3
  • Antigen name:
  • CD3 Molecule
  • CD3 Molecule
  • Antigen synonyms:
  • CD62L (L-Selectin)|CD44|CD3|CD8a|CD25|CD4
  • CD62L (L-Selectin)|CD44|CD3|CD8a|CD25|CD4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • 17A2, RM4-5, 53-6.7, PC61.5, IM7, MEL-14
  • 17A2, RM4-5, 53-6.7, PC61.5, IM7, MEL-14
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together