Anti-MME Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SN5c]
Supplier: Tonbo Biosciences
Certificates
About this item
The SN5c antibody is specific for human CD10, a 100 kDa type II transmembrane protein also known as Common Acute Lymphoblastic Leukemia Antigen (CALLA). In normal cells, it is expressed on early B and T lymphoid precursors, neutrophils and on various epithelia. CD10 is also expressed on several lymphoma cells including Burkitt's, acute lymphoblastic, and follicular germinal center lymphomas. CD10 functions as an endopeptidase and is involved in B cell development, as well as a mediator of neutrophil inflammatory responses.
Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: SN5c
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- 20-0108-T025
- 20-0108-T500
- 20-0108-T100
- Antigen symbol:
- MME
- MME
- MME
- Antigen name:
- Membrane metallo-endopeptidase
- Membrane metallo-endopeptidase
- Membrane metallo-endopeptidase
- Antigen synonyms:
- NEP|CD10|CALLA|SFE
- NEP|CD10|CALLA|SFE
- NEP|CD10|CALLA|SFE
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- SN5c
- SN5c
- SN5c
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 4311
- 4311
- 4311
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Yes
- Cross adsorption:
- No
- No
- No
- Storage temperature:
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.
Specifications
Frequently Bought Together