Anti-CD20 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2H7]
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation.
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
- Catalog No:
- 35-0209-T500
- 35-0209-T025
- 35-0209-T100
- Antigen symbol:
- MS4A1
- MS4A1
- MS4A1
- Antigen name:
- Membrane-spanning 4-Domains, Subfamily A, Member 1
- Membrane-spanning 4-Domains, Subfamily A, Member 1
- Membrane-spanning 4-Domains, Subfamily A, Member 1
- Antigen synonyms:
- CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
- CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
- CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 2H7
- 2H7
- 2H7
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 931
- 931
- 931
- Isotype:
- IgG2b kappa
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Yes
- Cross adsorption:
- No
- No
- No
- Storage temperature:
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.
Frequently Bought Together