Order Entry
Austria
ContactUsLinkComponent
 
Anti-CD20 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2H7]
 
Anti-CD20 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2H7]

 

  • Catalog No:
  • 20-0209-T500
  • 20-0209-T025
  • 20-0209-T100
  • Antigen symbol:
  • MS4A1
  • MS4A1
  • MS4A1
  • Antigen name:
  • Membrane-spanning 4-Domains, Subfamily A, Member 1
  • Membrane-spanning 4-Domains, Subfamily A, Member 1
  • Membrane-spanning 4-Domains, Subfamily A, Member 1
  • Antigen synonyms:
  • CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
  • CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
  • CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 2H7
  • 2H7
  • 2H7
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 931
  • 931
  • 931
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • IgG2b kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

 

Frequently Bought Together