Equivalence C18, HPLC Columns
Supplier: Avantor
Certificates
About this item
Stainless steel Equivalence C18 columns have been specifically developed as a high quality, cost effective replacement for many HPLC column brands.
- Excellent efficiency and reproducibility
- Available in 3 or 5 µm particle sizes
Maximum recommended pH: 2,0 to 8,0
Specifications
- Catalog No:
- EQV-5C18-1504
- EQV-5C18-1002
- EQV-3C18-1546
- EQV-3C18-1046
- EQV-3C18-0346
- EQV-3C18-0546
- EQV-5C18-2504
- EQV-5C18-2546
- EQV-3C18-GD2
- EQV-5C18-1246
- EQV-5C18-1203
- EQV-5C18-1046
- EQV-5C18-1003
- EQV-5C18-7546
- EQV-5C18-0546
- EQV-3C18-1203
- EQV-5C18-1546
- EQV-5C18-GD
- EQV-3C18-1246
- EQV-3C18-7546
- EQV-3C18-GD
- EQV-3C18-0502
- EQV-3C18-1503
- EQV-3C18-1002
- EQV-3C18-1502
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)