ContactUsLinkComponent
RNA loading dye (2X)
RNA loading dye (2X)
Catalog # R0641
CAS Number:  
undefined
RNA loading dye (2X)
Catalog # R0641
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    RNA loading dye 2X
  • Pk Info:
    1 ml
Specifications
About this item
In most denaturing agarose gel systems, bromophenol blue migrates slightly faster than human 5S rRNA, whereas xylene cyanol FF migrates slightly slower than 18S rRNA. RNA loading dye contains the denaturing agent formamide, which allows RNA fragments to separate according to size even during non-denaturing electrophoresis. Formamide also stabilises RNA.
  • Two-colour tracking of DNA, RNA migration during electrophoresis
  • No DNA, RNA masking during gel exposure to UV light
  • Denaturation of DNA, RNA due to the presence of formamide
Recommended for preparation of RNA samples for electrophoresis on agarose or polyacrylamide gels.
Ordering information: Contains the tracking dyes bromophenol blue and xylene cyanol FF, as well as the intercalating dye ethidium bromide.