HPLC columns, Hibar®, LiChrospher® 100 PAH
Supplier: Merck
Certificates
About this item
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.
Frequently Bought Together