Order Entry
Austria
ContactUsLinkComponent
VWR® Bottles, Wide Neck, Square, with Screw Caps
VWR® Bottles, Wide Neck, Square, with Screw Caps
Catalog # VWRLSTL101.1
New Product
undefined
VWR® Bottles, Wide Neck, Square, with Screw Caps
Catalog # VWRLSTL101.1
New Product

Specifications

  • Thread:
    38 mm
  • Capacity:
    500 ml
  • Description:
    unlabelled
  • Seq:
    03
  • W×D×H:
    63,50x63,50x184,0 mm
  • Environmentally Preferable:
  • Cap colour:
    White
  • Pk:
    120

Specifications

About this item

PET, clear, with screw cap.

  • Good chemical resistance
  • With or without label available

Ideal for packaging, storing and transporting liquids, pastes and powders. Wide mouth for easy filling, emptying and cleaning.