Orders LinkContactUsLinkComponent
Control Cables
Control Cables
Supplier:   HUBER
Control Cables
Supplier:  HUBER
Certificates
Control Cables
8 Options Available Below
Product Details & Documents
A range of control cables is available for USB, RS232, RS485 or analogue interfaces (AIF). Cables are also available for external control signal (ECS), a potentialfree contact (POKO) or for an external float switch (LEVEL).
Product Family Options
Product Information
  • Length
    Availability
    Price
  • HUBE54949
    Control cable to connect Pilot ONE®, Mini-USB to PC, USB Type A
    Control cable to connect Pilot ONE®, Mini-USB to PC, USB Type A
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,00 €
    17,00 €
    1 * 1 ST
       17,00 €
  • HUBE55018
    Control cable to connect units with RS232 (15 pol.) to PC, 9 pol. sub-D
    Control cable to connect units with RS232 (15 pol.) to PC, 9 pol. sub-D
    3 m
    Log in for availability
    Log in for availability
    1 * 3 m
    1 * 3 m - 155,00 €
    155,00 €
    1 * 3 m
       155,00 €
  • 461-0622
    Control cable to connect units with RS232 (9 pol.) / Com.G@te® to PC, 9 pol. sub-D
    Control cable to connect units with RS232 (9 pol.) / Com.G@te® to PC, 9 pol. sub-D
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 155,00 €
    155,00 €
    1 * 1 ST
       155,00 €
  • HUBE6279
    Control cable to connect Com.G@te® RS485 to cable end open
    Control cable to connect Com.G@te® RS485 to cable end open
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,00 €
    89,00 €
    1 * 1 ST
       89,00 €
  • HUBE9353
    Control cable to connect Com.G@te® AIF to cable end open
    Control cable to connect Com.G@te® AIF to cable end open
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,00 €
    89,00 €
    1 * 1 ST
       89,00 €
  • HUBE9490
    Control cable to connect Com.G@te® POKO to cable end open
    Control cable to connect Com.G@te® POKO to cable end open
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,00 €
    89,00 €
    1 * 1 ST
       89,00 €
  • HUBE9491
    Control cable to connect Com.G@te® ECS to cable end open
    Control cable to connect Com.G@te® ECS to cable end open
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,00 €
    89,00 €
    1 * 1 ST
       89,00 €
  • HUBE9492
    Control cable to connect Com.G@te® LEVEL to cable end open
    Control cable to connect Com.G@te® LEVEL to cable end open
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,00 €
    89,00 €
    1 * 1 ST
       89,00 €