Order Entry
Austria
ContactUsLinkComponent
Gas detection tubes for hydrogen fluoride
Gas detection tubes for hydrogen fluoride
Supplier:  Dräger
Certificates
undefined
Gas detection tubes for hydrogen fluoride
Supplier:  Dräger

Specifications

  • Catalog No:
  • 118-0321
  • 151-0113
  • Model:
  • Hydrogen fluoride 0.5/a
  • Hydrogen fluoride 1.5/b
  • Colour change:
  • Blue violet to yellow
  • Pale blue to pale pink
  • Humidity:
  • 30 - 80 % RH
  • ≤ 9 mg/l H₂O
  • Measurement time:
  • 2 minutes/25 seconds
  • 2 minutes
  • No. of strokes:
  • 10 / 2
  • 20
  • Standard deviation:
  • ±20 - 30 %
  • Standard range:
  • 0,5 - 15/10 - 90 ppm
  • 1,5 - 15 ppm
  • Temperature range:
  • 10…40 °C
  • 15…30 °C

Specifications

Frequently Bought Together