ContactUsLinkComponent
NBB®-P, powder
NBB®-P, powder
Catalog # 710-0743
Supplier:  DOEHLER GROUP
CAS Number:  
undefined
NBB®-P, powder
Catalog # 710-0743
Supplier:  DOEHLER GROUP
CAS Number:  
Specifications
  • Pk:
    300 g
  • Description:
    NBB®-powder (NBB®-P)
Specifications
About this item
Powder for production of NBB®-Broth and NBB®-Agar for the detection of Beer spoiling bacteria in filtered beer samples, water samples, yeast samples as well as for membrane and swab samples. Under its NBB® brand, Döhler offers innovative culture media for the fast and reliable detection of beverage spoiling microorganisms, such as Lactobacilli, Pectinatus and Megasphaera, in all samples that arise in breweries, as well as for the wine industry. NBB®-Powder (NBB®-P) is a powder, which dissolved in your own beer, will adapt the selectivity of NBB®-Media onto the demand of your products. NBB® culture media are suitable for detection in:
  • Yeast samples
  • Unfiltered beers containing yeast
  • Filtered beers
  • Wine
  • New wine
  • Water and rinsing water
  • Airborne bacteria collection samples
  • Swab samples for hygiene monitoring