Order Entry
Austria
ContactUsLinkComponent
VWR® Storage/Media Bottles with or without Screw Cap
VWR® Storage/Media Bottles with or without Screw Cap
Catalog # 213-0429
Supplier:  VWR Collection
undefined
VWR® Storage/Media Bottles with or without Screw Cap
Catalog # 213-0429
Supplier:  VWR Collection

Specifications

  • Thread:
    GL 45
  • Capacity:
    2000 ml
  • Seq:
    05
  • Neck I-Ø:
    33 mm
  • Pk:
    10

Specifications

About this item

Borosilicate glass, clear.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

VWR® storage/media bottles are designed with a wide opening and a drip-free polypropylene pouring ring. They feature enhanced graduations and marking spots printed in permanent white enamel.