Trays, bare die/CSP, H 44 Series
Supplier: ENTEGRIS
Certificates
About this item
Entegris offers a wide selection of H 44 series 4" square trays, covers and accessories to meet your changing needs. H 20 series trays are available in various pocket sizes and depths, with or without wash holes and tweezers slots. H 44 series products offer proven reliability for handling bare die, including:
- Known good die (KGD)
- Wafer scale packing (WSP)
- Chip scal packing (CSP)
- Flip chip
- Bump die
- Opto electronics
- Passive and active chip components, mechanical components
Frequently Bought Together