ContactUsLinkComponent
Gas detection tubes for carbon monoxide
Gas detection tubes for carbon monoxide
Supplier:  Dräger
Certificates
undefined
Gas detection tubes for carbon monoxide
Supplier:  Dräger
Certificates
Specifications
  • Catalog No:
  • 151-3159
  • 151-0190
  • 118-0347
  • 151-0188
  • 151-0162
  • Model:
  • Carbon monoxide 5/c
  • Carbon monoxide 10/b
  • Carbon monoxide 0.3%/b
  • Carbon monoxide 8/a
  • Carbon monoxide 2/a
  • Colour change:
  • White to brownish green
  • White to brownish green
  • White to brownish green
  • White to pale brown
  • White to brownish
  • Measurement time:
  • 50 seconds/2,5 minutes
  • 20 seconds/4 minutes
  • 30 seconds
  • 2 minutes
  • 4 minutes/50 seconds
  • No. of strokes:
  • 1/5
  • 1/10
  • 1
  • 10
  • 10/2
  • Standard deviation:
  • ±10 - 15 %
  • ±10 - 15 %
  • ±10 - 15 %
  • ±10 - 15 %
  • ±10 - 15 %
  • Standard range:
  • 100 - 700/5 - 150 ppm
  • 100 - 3000/10 - 300 ppm
  • 0,3 - 7 vol %  
  • 8 - 150 ppm
  • 2 - 60/25 - 300 ppm