Safety wash bottles, narrow neck, right-to-understand, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
LDPE, translucent.
- Self-venting wash bottles eliminate risk of bottles self-dispensing
- Good chemical resistance
- Good temperature resistance
Prevent chemical accidents in the lab with Right-to-Understand wash bottles. Bottles are pre-labeled with chemical identification and safe handling information following the Globally Harmonized System (GHS) specifications as applicable to secondary laboratory containers. Wash bottles are ready to use when you transfer common laboratory chemicals from manufacturers’ containers.
These cylindrical bottles are not autoclavable and not graduated.
Specifications
- Catalog No:
- 215-3830
- 215-3829
- 215-3833
- 215-3834
- 215-3831
- 215-3832
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement