Order Entry
Austria
ContactUsLinkComponent
 
VWR®, Reagent reservoirs
  89094-664
undefined
VWR®, Reagent reservoirs
  89094-664

 

  • Packed:
    Individually wrapped
  • Sterile:
    +
  • Type:
    Standard
  • Capacity:
    25 ml
  • Colour:
    White
  • Seq:
    01
  • W×D×H:
    146×51×25 mm
  • Pk:
    100

 

 

PS, sterile or non sterile. These disposable pipetting reservoirs are designed for use in multi channel pipetting applications, and will accommodate all types of pipettors up to 12-channel width.

  • Lot-certified, non pyrogenic, DNase- and RNase-free
  • Trough design, graduation marks on interior walls, and pour-off spouts on each corner
  • 25 ml reservoirs are also available in a unique divided format that features three distinct mini reservoir chambers, each with a 5 ml capacity
 : Available bulk packed or individually wrapped.