ContactUsLinkComponent
Peqlab peqGOLD, DNA Ladder, 1 kb
Peqlab peqGOLD, DNA Ladder, 1 kb
Catalog # 25-2030
Supplier:  VWR Chemicals
CAS Number:  
undefined
Peqlab peqGOLD, DNA Ladder, 1 kb
Catalog # 25-2030
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    DNA ladder, 1 kb (concentrated), 500 ng/µl, 5×100 µl, sufficient for 250 lanes
Specifications
About this item
DNA ladder for quantitative and qualitative analysis of dsDNA.
  • Range: 14 fragments, 250 to 10000 bp in size
  • Three highlighted bands for easy orientation
  • Also available as ready to use mix containing Orange G dye instead of bromophenol blue