Orders LinkContactUsLinkComponent
Hydrometer set
Hydrometer set
Complete set of 14 instruments, 1
Catalog # AMARH809630
Supplier:   Amarell
Hydrometer set
Catalog # AMARH809630
Supplier:  Amarell
Hydrometer set
Complete set of 14 instruments, 1
Price
Inquire For Price
Log in for availability
1 * 1 SET
Product Details & Documents
Hydrometers with scale paper, colour code and type designation, weighted body, with (and without) thermometer inside the hydrometer body, round capillary, mercury filling suitable for government verification.
Delivery information: Supplied in a transparent plastic case.
Specifications
  • Description:
    Complete set of 14 instruments
  • Division:
    1