VWR®, Labelling Tapes, General Purpose Laboratory
VWR® general-purpose laboratory labelling tape sticks to virtually any surface, can be marked with most writing utensils, and reliably withstands extreme temperature variations. It can be written on with pencil, fiber-tip or ball point pen, and lab markers.
- Vinyl-coated soft satin-finish paper
- Adhesive sticks to virtually any surface
- Tape removal leaves no residue
This durable self-adhesive tape is sutiable for general laboratory use and is excellent for applications in refrigeration. It can withstand temperatures from –23 to 121 °C (–10 to 250 °F) and has an application temperature as low as 0 °C (32 °F). The adhesive backing will peel off easily and leaves no residue.
Length: 12,7 and 54,8 m
Frequently Bought Together