ContactUsLinkComponent
Peqlab peqGOLD, DNA Ladders, 50 bp
Peqlab peqGOLD, DNA Ladders, 50 bp
Catalog # 25-2000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Peqlab peqGOLD, DNA Ladders, 50 bp
Catalog # 25-2000
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    DNA ladder, 50 bp (concentrated), 500 ng/µl, 1×100 µl, sufficient for 50 lanes
Specifications
About this item
DNA ladder for quantitative and qualitative analysis of dsDNA.
  • Range: 13 distinct fragments 50 to 1000 bp in size
  • Two highlighted bands for easy orientation
  • Also available as ready to use mix containing Orange G dye instead of bromophenol blue