Orders LinkContactUsLinkComponent
Digital interval timer
Digital interval timer
Digital interval timer, 99 min, 59 s, White
Catalog # HECH41977020
Digital interval timer
Catalog # HECH41977020
Digital interval timer
Digital interval timer
Digital interval timer, 99 min, 59 s, White
Price
17,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Digital interval timer with white casing and magnetic back.
  • LCD display, 4 digits showing minutes and seconds
  • Programmable up to 99 minutes and 59 seconds
  • Counts down in seconds
  • Alarm sounds for 30 seconds when countdown reaches zero
  • Memory recall function
  • Individual setting for minutes and seconds
Specifications
  • Colour:
    White
  • Description:
    Digital interval timer, 99 min, 59 s