ContactUsLinkComponent
Vacuum trap assembly, A584
Vacuum trap assembly, A584
Catalog # VARIA584
Supplier:  VARIAN
Vacuum trap assembly, A584
Catalog # VARIA584
Supplier:  VARIAN
Specifications
  • Description:
    Vacuum trap assembly, includes 1 glass jar, 1 stopper, 2 elbow fittings and 2 tubes (I.Ø 6,35 mm, O.Ø 11,1 mm, length 457 mm)
  • Pk:
    1
Specifications
About this item
This vacuum trap eliminates the possibility of drawing fluids into the vacuum pump that may be detrimental to it.