Order Entry
Austria
ContactUsLinkComponent
Quartz microfibre filters, pre-heated, grade Q3400
Quartz microfibre filters, pre-heated, grade Q3400
Supplier:  Sartorius
Certificates
undefined
Quartz microfibre filters, pre-heated, grade Q3400
Supplier:  Sartorius

Specifications

  • Catalog No:
  • SARTQ3400-90-N
  • SARTQ3400-20-G
  • SARTQ3400-82-N
  • SARTQ3400-150-K
  • SARTQ3400-142-K
  • SARTQ3400-47-G
  • SARTQ3400-45-G
  • SARTQ3400-50-G
  • SARTQ3400-37-G
  • SARTQ3400-25-G
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 0,43 mm
  • 0,43 mm
  • 0,43 mm
  • 0,43 mm
  • 0,43 mm
  • 0,43 mm
  • 0,43 mm
  • 0,43 mm
  • 0,43 mm
  • 0,43 mm
  • Particle retention:
  • 99,998%
  • 99,998%
  • 99,998%
  • 99,998%
  • 99,998%
  • 99,998%
  • 99,998%
  • 99,998%
  • 99,998%
  • 99,998%
  • Max. temperature:
  • 900 °C
  • 900 °C
  • 900 °C
  • 900 °C
  • 900 °C
  • 900 °C
  • 900 °C
  • 900 °C
  • 900 °C
  • 900 °C

Specifications

Frequently Bought Together