Order Entry
Austria
ContactUsLinkComponent
Wafer storage boxes for 2,5" and smaller wafer handling, E 04 series
Wafer storage boxes for 2,5" and smaller wafer handling, E 04 series
Catalog # ENTEE04-1202
Supplier:  ENTEGRIS
Wafer storage boxes for 2,5" and smaller wafer handling, E 04 series
Catalog # ENTEE04-1202
Supplier:  ENTEGRIS

Specifications

  • Colour:
    Black
  • Material:
    Polystyrene
  • Pk:
    4

Specifications

About this item

  • Textured, nonslip surface
  • Liftoff cover
  • Friction seal only
  • Box capacity: one carrier
  • Compatible carriers: 2", 2,25" and 2,5" A72 and PA72 wafer carriers