Order Entry
Austria
ContactUsLinkComponent
Multiple wafer shipping boxes for 76,2 mm (3") wafer handling, H 6630 series
Multiple wafer shipping boxes for 76,2 mm (3") wafer handling, H 6630 series
  ENTEH6630-0009_U
 :  ENTEGRIS
Multiple wafer shipping boxes for 76,2 mm (3") wafer handling, H 6630 series
  ENTEH6630-0009_U
 :  ENTEGRIS

 

  • Description:
    Assembly, shrink-wrapped, PP, natural
  • Pk:
    60

 

 

  • Full-depth wafer protection
  • Positive cover alignment
  • Individual wafer support springs
  • Stackable
  • Assembly overall size: 160×60×67 mm
  • Pitch: 4,76 mm