Order Entry
Austria
ContactUsLinkComponent
Wafer carrier handles for 76,2 mm (3") wafer handling
Wafer carrier handles for 76,2 mm (3") wafer handling
Catalog # ENTEA82-03-0215
Supplier:  ENTEGRIS
Wafer carrier handles for 76,2 mm (3") wafer handling
Catalog # ENTEA82-03-0215
Supplier:  ENTEGRIS

Specifications

  • For:
    A82M, PA82M wafer carriers
  • Colour:
    Natural
  • Description:
    Squeeze
  • Material:
    PFA
  • Pk:
    4

Specifications

About this item

  • Attaches to the notches on both end walls of carrier
  • Overall size: 165×91×157 mm