Orders LinkContactUsLinkComponent
Flame atomisation systems
Flame atomisation systems
Supplier:   VARIAN
Flame atomisation systems
Supplier:  VARIAN
Certificates
Flame atomisation systems
11 Options Available Below
Product Details & Documents
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
  • Contoured design for outstanding resistance to blockage even with the most difficult examples
  • Corrosion-resistant components
  • For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
Ordering information: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.
Product Family Options
Product Information
  • Availability
    Price
  • VARI190034100
    Flame atomisation system kit
    Flame atomisation system kit
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 976,93 €
    2 976,93 €
    1 * 1 ST
       2 976,93 €
Spray chamber assembly
Product Information
  • Availability
    Price
  • VARI110634490
    Complete spray chamber assembly for aqueous solutions fitted with O-rings
    Complete spray chamber assembly for aqueous solutions fitted with O-rings
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 263,37 €
    3 263,37 €
    1 * 1 ST
       3 263,37 €
  • VARI110637000
    Mixing paddle, fluorinated
    Mixing paddle, fluorinated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,25 €
    60,25 €
    1 * 1 ST
       60,25 €
  • VARI9910053300
    PTFE impact beads for HF digests
    PTFE impact beads for HF digests
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 977,99 €
    977,99 €
    1 * 1 ST
       977,99 €
Nebuliser components
Product Information
  • Availability
    Price
  • VARI1610117390
    PEEK venturi nebuliser
    PEEK venturi nebuliser
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 260,86 €
    260,86 €
    1 * 1 ST
       260,86 €
  • VARI9910024700
    Cleaning wire (Lר = 3×0,3 m)
    Cleaning wire (Lר = 3×0,3 m)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,88 €
    65,88 €
    1 * 1 ST
       65,88 €
  • VARI9910043700
    High vacuum preset barrel nebuliser
    High vacuum preset barrel nebuliser
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 882,32 €
    1 882,32 €
    1 * 1 ST
       1 882,32 €
  • VARI9910044000
    High vacuum capillary tubing
    High vacuum capillary tubing
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,42 €
    45,42 €
    1 * 1 ST
       45,42 €
Burners
Product Information
  • Availability
    Price
  • VARI210164000
    Air/acetylene burner assembly
    Air/acetylene burner assembly
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 964,16 €
    1 964,16 €
    1 * 1 ST
       1 964,16 €
  • VARI210164100
    Nitrous oxide/acetylene burner assembly
    Nitrous oxide/acetylene burner assembly
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 964,16 €
    1 964,16 €
    1 * 1 ST
       1 964,16 €
  • VARI9910053900
    Burner cleaning and alignment card assembly
    Burner cleaning and alignment card assembly
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,13 €
    59,13 €
    1 * 1 ST
       59,13 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"
    Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"
    VARI6910009700
    Log in for availability
    1 * 1 ST - 23,94 €
Product Information
  • Catalog #
    Availability
    Price
  • Glass impact beads
    Glass impact beads
    VARI9910025700
    Log in for availability
    1 * 5 ST - 210,74 €
Product Information
  • Catalog #
    Availability
    Price
  • Aqueous spray chamber o-ring
    Aqueous spray chamber o-ring
    VARI9910093400
    Log in for availability
    1 * 1 ST - 89,72 €
Product Information
  • Catalog #
    Availability
    Price
  • Organics o-ring
    Organics o-ring
    VARI9910093500
    Log in for availability
    1 * 1 ST - 1 207,14 €
Product Information
  • Catalog #
    Availability
    Price
  • Fluorinated mixing paddles
    Fluorinated mixing paddles
    VARI9910093600
    Log in for availability
    1 * 1 ST - 260,86 €
Product Information
  • Catalog #
    Availability
    Price
  • Integrated liquid trap (fluorinated float and magnet)
    Integrated liquid trap (fluorinated float and magnet)
    VARI9910105800
    Log in for availability
    1 * 1 ST - 439,89 €
Product Information
  • Catalog #
    Availability
    Price
  • Nebuliser capillary kit
    Nebuliser capillary kit
    VARI9910093000
    Log in for availability
    1 * 1 ST - 790,78 €
Product Information
  • Catalog #
    Availability
    Price
  • Integrated nebuliser block
    Integrated nebuliser block
    VARI9910093100
    Log in for availability
    1 * 1 ST - 1 401,51 €
Product Information
  • Catalog #
    Availability
    Price
  • Flame atomisation system kit
    Flame atomisation system kit
    VARI190034100
    Log in for availability
    1 * 1 ST - 2 976,93 €