ContactUsLinkComponent
Manually operated diaphragm valves, Integra®
Manually operated diaphragm valves, Integra®
Catalog # ENTE201-42-01-3
Supplier:  ENTEGRIS
Manually operated diaphragm valves, Integra®
Catalog # ENTE201-42-01-3
Supplier:  ENTEGRIS
Specifications
  • Description:
    PFA, port connection 1/2" Flaretek® flared tube
  • Pk:
    1
Specifications
About this item
Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution systems. Three and one-half complete turns fully opens or closes the valve for more precise control than a ¹/₄-turn valve. Slower actuation reduces water hammer affect on fluid stream. All wetted parts are molded PFA providing chemical resistance suitable for use in high purity, high corrosion chemical handling applications.
  • No external metal components
  • Red travel indicator allows quick visual determination of diaphragm position
  • Field serviceable
  • Addition of Espy® sensor allows electronic monitoring of valve body seal integrity
¹/₂" orifice, 2-way, multi-turn, leak detection option.