ContactUsLinkComponent
VWR®, Cryoboxes
VWR®, Cryoboxes
Catalog # 478-0241
Supplier:  VWR Collection
undefined
VWR®, Cryoboxes
Catalog # 478-0241
Supplier:  VWR Collection
Specifications
  • Compartments:
    4×4
  • Height:
    - mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Polycarbonate.
  • Can be used from –190 to +121 °C
VWR® freezer boxes can be used with most manufacturer’s upright and chest freezer racks in general purpose and ultra-low mechanical freezers.