ContactUsLinkComponent
gelStar™ Nucleic acid gel stain 10,000X
Catalog # LONZ50535
Supplier:  LONZA
CAS Number:  
undefined
gelStar™ Nucleic acid gel stain 10,000X
Catalog # LONZ50535
Supplier:  LONZA
CAS Number:  
Specifications
  • Pk:
    500 µl
  • Description:
    GelStar™ Nucleic Acid Gel Stain 10,000X
  • Pk Info:
    2 x 250 µl
Specifications
About this item
GelStar is a sensitive fluorescent stain for detecting both DNA and RNA.
  • Maximum sensitivity - detect as little as 20 pg of dsDNA or 3 ng of RNA,
  • Versatile - use for agarose or polyacrylamide gel electrophoresis,
  • Ideal alternative to silver staining
GelStar™ Nucleic Acid Gel Stain is a highly sensitive fluorescent stain for detecting both DNA and RNA. You can choose to either add GelStarTM Stain to your agarose solution prior to casting or to post stain your gels. GelStarTM Nucleic Acid Gel Stain exhibits exceptional signal-to-noise ratio with minimal background. Maximum sensitivity, detect as little as 20 pg of dsDNA or 3 ng of RNA