Order Entry
Austria
ContactUsLinkComponent
Skeleton model, with hanging stand
Skeleton model, with hanging stand
Catalog # BINH1020172
Supplier:  3B Scientific
undefined
Skeleton model, with hanging stand
Catalog # BINH1020172
Supplier:  3B Scientific

Specifications

  • Description:
    Skeleton model - Stan with hanging stand, 192,5 mm
  • Pk:
    1

Specifications

About this item

Stan, the standard model of a human skeleton has a very high quality and robust construction along with a stable metal hanging stand.

  • Made from a durable, unbreakable synthetic material
  • Hanging stand with five casters (painted white)
  • 3 part assembled skull and individually inserted teeth
  • Limbs can be removed quickly and easily

Weight: 8,77 kg