ContactUsLinkComponent
Agarose, SeaPrep™
Agarose, SeaPrep™
  LONZ50302
 :  LONZA
Agarose, SeaPrep™
  LONZ50302
 :  LONZA
  • Description:
    Agarose, SeaPrep™
  • Pk:
    25 g
Ultra-soft agarose, ideal for high efficiency hybridoma cloning and expansion of cDNA libraries.