VWR®, Culture Tubes with Black PP Screw Cap
Supplier: VWR Collection
Certificates
About this item
Made from soda-lime glass, clear.
- GL thread
- Wall thickness 1 mm (±0,04 mm)
- High chemical and mechanical resistance
Caution: Note: Caps are not autoclavable.