Orders LinkContactUsLinkComponent
Test charts
Test charts
Supplier:   Industrial Physics
Test charts
Supplier:  Industrial Physics
Certificates
Test charts
21 Options Available Below
Product Details & Documents
Test charts are used for testing physical properties of coating, lacquers, and inks.
  • Dimensions from A6 up to A4
  • Film laminated for an excellent solvent, chemical resistance, and film spread
  • Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Ordering information: A certificate is available for charts with an optical brightener.
Product Family Options
Product Information
  • Colour
    Format
    W×H
    Availability
    Price
  • SIMEVF2317
    Test charts
    Test charts
    White/black
    A6
    105×148 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 88,70 €
    88,70 €
    1 * 250 ST
       88,70 €
  • SIMEVF2343
    Test charts with optical brightner
    Test charts with optical brightner
    White/black
    A6
    105×148 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 80,10 €
    80,10 €
    1 * 250 ST
       80,10 €
  • SIMEVF2319
    Test charts
    Test charts
    White/black
    A5
    148×210 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 138,00 €
    138,00 €
    1 * 250 ST
       138,00 €
  • SIMEVF2344
    Test charts with optical brightner
    Test charts with optical brightner
    White/black
    A5
    148×210 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 113,00 €
    113,00 €
    1 * 250 ST
       113,00 €
  • SIMEVF2323
    Test charts
    Test charts
    Black/white chequered
    A5
    148×210 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 158,00 €
    158,00 €
    1 * 250 ST
       158,00 €
  • SIMEVF2346
    Test charts with optical brightner
    Test charts with optical brightner
    Black/white chequered
    A5
    148×210 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 120,00 €
    120,00 €
    1 * 250 ST
       120,00 €
  • SIMEVF2321
    Test charts
    Test charts
    White/black
    A4
    210×297 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 220,00 €
    220,00 €
    1 * 250 ST
       220,00 €
  • SIMEVF2345
    Test charts with optical brightner
    Test charts with optical brightner
    White/black
    A4
    210×297 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 157,00 €
    157,00 €
    1 * 250 ST
       157,00 €
  • SIMEVF2325
    Test charts
    Test charts
    Black/white chequered
    A4
    210×297 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 278,00 €
    278,00 €
    1 * 250 ST
       278,00 €
  • SIMEVF2347
    Test charts with optical brightner
    Test charts with optical brightner
    Black/white chequered
    A4
    210×297 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 184,00 €
    184,00 €
    1 * 250 ST
       184,00 €
  • SIMEVF2354
    Test charts with optical brightner
    Test charts with optical brightner
    Black/white chequered
    A3
    297×420 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 285,00 €
    285,00 €
    1 * 250 ST
       285,00 €
  • SIMEVF2316
    Opacity scrub charts with optical brightner
    Opacity scrub charts with optical brightner
    White/black
    165×480 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 285,00 €
    285,00 €
    1 * 100 ST
       285,00 €
Leneta test charts
Product Information
  • Colour
    Format
    W×H
    Availability
    Price
  • SIMEVF3001
    Test charts
    Test charts
    White/black
    8B
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 314,00 €
    314,00 €
    1 * 250 ST
       314,00 €
  • SIMEVF3000
    Test charts
    Test charts
    White/black
    2C
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 252,00 €
    252,00 €
    1 * 250 ST
       252,00 €
  • SIMEVF3006
    Test charts
    Test charts
    White/black
    5C
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 231,00 €
    231,00 €
    1 * 250 ST
       231,00 €
  • SIMEVF3003
    Test charts
    Test charts
    White/black
    2DX
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 184,00 €
    184,00 €
    1 * 500 ST
       184,00 €
  • SIMEVF3002
    Test charts
    Test charts
    White/black
    6F6
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 242,00 €
    242,00 €
    1 * 500 ST
       242,00 €
  • SIMEVF3007
    Test charts
    Test charts
    White/black
    12G
    Log in for availability
    Log in for availability
    1 * 125 ST
    1 * 125 ST - 495,00 €
    495,00 €
    1 * 125 ST
       495,00 €
  • SIMEVF3005
    Test charts
    Test charts
    White/black
    M12BW
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 460,00 €
    460,00 €
    1 * 2.000 ST
       460,00 €
  • SIMEVF3004
    Test charts
    Test charts
    White/black
    P121-10N
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 648,00 €
    648,00 €
    1 * 100 ST
       648,00 €
  • SIMEVF3008
    Test charts
    Test charts
    White
    WDX
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 226,00 €
    226,00 €
    1 * 500 ST
       226,00 €