Hydrometers for sodium chloride
Supplier: Ludwig Schneider
Jump to:
About this item
Made of high quality plastics, these hydrometers are designed for measurements in salt solutions.
- Breakage protected (no glass breakage)
- High transparency with easy-to-read scales
- High precision and reproducibility
Suitable for applications in food, pharmaceutical and cosmetic industries.
Ordering information: A works calibration certificate is available on request.
Packaging: In a square plastic sleeve.
2 Options Available Below
Product Details & Documents
Made of high quality plastics, these hydrometers are designed for measurements in salt solutions.
- Breakage protected (no glass breakage)
- High transparency with easy-to-read scales
- High precision and reproducibility
Suitable for applications in food, pharmaceutical and cosmetic industries.
Ordering information: A works calibration certificate is available on request.
Packaging: In a square plastic sleeve.