Traps for turbomatrix thermal desorbers
Lieferant: PerkinElmer
Zertifikate
Über diesen Artikel
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Lieferant: VWR Chemicals
Acetone ≥99%, TECHNICAL
Lieferant: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Lieferant: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Lieferant: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Lieferant: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Lieferant: Merck
VWR®, Crimp Neck Vials ND11
Lieferant: VWR Collection
VWR® Facial Tissues
Lieferant: VWR Collection
VWR®, Nitrile Gloves
Lieferant: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Lieferant: EPPENDORF
VWR®, Short Thread Vials ND9
Lieferant: VWR Collection
VWR®, Transfer Pipettes, High Performance
Lieferant: VWR Collection
Gas-tight syringes, 10 to 500 µl
Lieferant: Trajan Scientific and Medical
FILTER -TBMTX TD COATED SS DISK 1 * 1 items
Lieferant: PerkinElmer