ContactUsLinkComponent
Alcometer, recalibration
Alcometer, recalibration
Catalog # SUREAMCAL
Supplier:  SURESCREEN
undefined
Alcometer, recalibration
Catalog # SUREAMCAL
Supplier:  SURESCREEN
Specifications
  • Description:
    Recalibration service for Alcometer breathalyser
  • Pk:
    1
Specifications
About this item
After 500 uses the alcometer requires recalibration to ensure that the instrument is reading correctly. It is returned to manufacturer for this service, and turn around is five working days from receipt.