Orders LinkContactUsLinkComponent
Mini hand press, MHP-1
Mini hand press, MHP-1
Mini hand press, MHP-1
Catalog # SHIM200-66747-91
Supplier:   SHIMADZU
Mini hand press, MHP-1
Catalog # SHIM200-66747-91
Supplier:  SHIMADZU
Mini hand press, MHP-1
Mini hand press, MHP-1
Price
2 270,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This compact mini hand press is a used to produce KBr pellets.
  • Pellet produced in the frame directly measured using the dedicated holder
  • For regular Ø 13 mm pellets, evacuable die, hydraulic press and vacuum pump are used
  • No dies or vacuum pump are required
Ordering information: Includes mini hand press, pellet holder, upper and lower cylinder, briquetting frame (Ø 4 mm), pellet remover and pellet remover base.
Specifications
  • Model:
    MHP-1
  • Description:
    Mini hand press
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Evacuable die
    Evacuable die
    SHIM202-32010
    Log in for availability
    1 * 1 ST - 1 480,00 €