Order Entry
Austria
ContactUsLinkComponent
Flexible brushes, nylon
Flexible brushes, nylon
Catalog # JMBR4470/0045
Supplier:  JM BROSSERIE
Flexible brushes, nylon
Catalog # JMBR4470/0045
Supplier:  JM BROSSERIE

Specifications

  • Bristle material:
    Nylon
  • Brush head Ø:
    32 mm
  • Description:
    Flexible brush, with rubber coated handle
  • Seq:
    01
  • Total length:
    450 mm
  • Brush head length:
    190 mm
  • Pk:
    1

Specifications

About this item

Flexible brushes made of nylon hair with rubber coated handle, a sheath of polyvinyl chloride.