Heat sealing foil, PeelASeal DMSO Foil™
Supplier: iST Scientific Ltd
Certificates
About this item
Peelable DMSO resistant foil for polypropylene plates. Suitable for low temperature and ambient temperature storage with DMSO and other solvents.
- Peelable
- Non pierceable
- High solvent resistance with good liquid barrier
- Seal integrity range: −80 to 80 °C